Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TFB1M
Entrez Gene ID
533438
Gene Name
transcription factor B1, mitochondrial
Gene Aliases
-
Chromosome Location
Chr.9: 91865417 - 91932613 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076896.2 NP_001070364.1 3 - 4 454 128
NM_001114610.1 NP_001108082.1 3 - 4 454 128
GenBank mRNA BC109841.1 - 3 - 4 442 128
DT811342.1 - 3 - 4 447 128
DY454254.1 - 3 - 4 458 128

View Full Product Details