Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C23H6orf62
Entrez Gene ID
534366
Gene Name
chromosome 23 C6orf62 homolog
Gene Aliases
-
Chromosome Location
Chr.23: 33054028 - 33068000 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077999.2 NP_001071467.1 4 - 5 1047 76
GenBank mRNA BC126615.1 - 4 - 5 659 76
BC148131.1 - 4 - 5 1939 76
BT030562.1 - 4 - 5 1047 76

View Full Product Details