Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RIT1
Entrez Gene ID
533646
Gene Name
Ras like without CAAX 1
Gene Aliases
-
Chromosome Location
Chr.3: 14853319 - 14862518 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098078.1 NP_001091547.1 1 - 2 53 82
GenBank mRNA BC140484.1 - 1 - 2 53 82

View Full Product Details