Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ESM1
Entrez Gene ID
539571
Gene Name
endothelial cell specific molecule 1
Gene Aliases
-
Chromosome Location
Chr.20: 24108178 - 24117012 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098101.1 NP_001091570.1 2 - 3 528 67
GenBank mRNA BC140693.1 - 2 - 3 528 67

View Full Product Details