Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EXOC8
Entrez Gene ID
540237
Gene Name
exocyst complex component 8
Gene Aliases
-
Chromosome Location
Chr.28: 4062688 - 4065152 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098112.2 NP_001091581.1 1 - 1 2345 146
GenBank mRNA BC134458.1 - 1 - 1 2347 146

View Full Product Details