Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KRT4
Entrez Gene ID
527819
Gene Name
keratin 4
Gene Aliases
-
Chromosome Location
Chr.5: 27117923 - 27124834 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098385.1 NP_001091855.1 4 - 5 920 72
GenBank mRNA BC134686.1 - 4 - 5 920 72

View Full Product Details