Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TNFRSF4
Entrez Gene ID
528782
Gene Name
TNF receptor superfamily member 4
Gene Aliases
-
Chromosome Location
Chr.16: 51285833 - 51288677 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099043.2 NP_001092513.1 2 - 3 302 73
GenBank mRNA BC142115.1 - 2 - 3 303 73

View Full Product Details