Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SERPINA11
Entrez Gene ID
538280
Gene Name
serpin family A member 11
Gene Aliases
-
Chromosome Location
Chr.21: 59147788 - 59161574 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099083.1 NP_001092553.1 4 - 5 1132 64
GenBank mRNA BC142389.1 - 4 - 5 1132 64

View Full Product Details