Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB6B
Entrez Gene ID
526526
Gene Name
RAB6B, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.1: 135468728 - 135526457 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101129.1 NP_001094599.1 2 - 3 249 89
GenBank mRNA BC150111.1 - 2 - 3 249 89

View Full Product Details