Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAT2A
Entrez Gene ID
527403
Gene Name
methionine adenosyltransferase 2A
Gene Aliases
-
Chromosome Location
Chr.11: 49465580 - 49472218 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101131.1 NP_001094601.1 5 - 6 658 63
GenBank mRNA BC111121.1 - 5 - 6 669 63
BT030708.1 - 5 - 6 658 63

View Full Product Details