Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASB16
Entrez Gene ID
539253
Gene Name
ankyrin repeat and SOCS box containing 16
Gene Aliases
-
Chromosome Location
Chr.19: 43989607 - 43997145 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101188.1 NP_001094658.1 2 - 3 646 105
GenBank mRNA BC147924.1 - 2 - 3 646 105

View Full Product Details