Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC39A9
Entrez Gene ID
539399
Gene Name
solute carrier family 39 member 9
Gene Aliases
-
Chromosome Location
Chr.10: 81185412 - 81230624 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101191.1 NP_001094661.1 5 - 6 1235 109
GenBank mRNA BC148060.1 - 5 - 6 1235 109

View Full Product Details