Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CRHBP
Entrez Gene ID
540087
Gene Name
corticotropin releasing hormone binding protein
Gene Aliases
-
Chromosome Location
Chr.10: 8104261 - 8118310 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101202.2 NP_001094672.1 4 - 5 643 76
GenBank mRNA BC151446.1 - 4 - 5 644 76

View Full Product Details