Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C22H3orf18
Entrez Gene ID
616712
Gene Name
chromosome 22 C3orf18 homolog
Gene Aliases
-
Chromosome Location
Chr.22: 49790596 - 49799041 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101260.1 NP_001094730.1 2 - 3 635 70
GenBank mRNA BC147991.1 - 2 - 3 635 70

View Full Product Details