Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM72A
Entrez Gene ID
616965
Gene Name
family with sequence similarity 72 member A
Gene Aliases
-
Chromosome Location
Chr.16: 3986148 - 3999330 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101263.1 NP_001094733.1 2 - 3 786 132
GenBank mRNA BC146229.1 - 2 - 3 786 132

View Full Product Details