Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VPS51
Entrez Gene ID
525567
Gene Name
VPS51 subunit of GARP complex
Gene Aliases
C29H11orf2, FFR
Chromosome Location
Chr.29: 43305622 - 43317745 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102093.2 NP_001095563.1 4 - 5 765 74
GenBank mRNA BC149640.1 - 4 - 5 766 74

View Full Product Details