Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC85A
Entrez Gene ID
525800
Gene Name
coiled-coil domain containing 85A
Gene Aliases
-
Chromosome Location
Chr.11: 38849960 - 39079710 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102095.1 NP_001095565.1 3 - 4 1553 67
GenBank mRNA BC148154.1 - 3 - 4 1553 67

View Full Product Details