Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DRC3
Entrez Gene ID
527539
Gene Name
dynein regulatory complex subunit 3
Gene Aliases
LRRC48
Chromosome Location
Chr.19: 34536472 - 34556492 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102111.1 NP_001095581.1 9 - 10 1440 77
GenBank mRNA BC149900.1 - 9 - 10 1440 77

View Full Product Details