Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYLIP
Entrez Gene ID
541070
Gene Name
myosin regulatory light chain interacting protein
Gene Aliases
-
Chromosome Location
Chr.23: 40992081 - 41012855 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102253.1 NP_001095723.1 3 - 4 568 63
GenBank mRNA BC149770.1 - 3 - 4 568 63

View Full Product Details