Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYRFL
Entrez Gene ID
781109
Gene Name
myelin regulatory factor like
Gene Aliases
C5H12orf28
Chromosome Location
Chr.5: 43516692 - 43642720 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102323.1 NP_001095793.1 12 - 13 1686 115
GenBank mRNA BC148934.1 - 12 - 13 1686 115

View Full Product Details