Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MYRFL
Entrez Gene ID
781109
Gene Name
myelin regulatory factor like
Gene Aliases
C5H12orf28
Chromosome Location
Chr.5: 43516692 - 43642720 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102323.1 NP_001095793.1 3 - 4 410 80
GenBank mRNA BC148934.1 - 3 - 4 410 80

View Full Product Details