Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NOP9
Entrez Gene ID
528833
Gene Name
NOP9 nucleolar protein
Gene Aliases
C10H14orf21
Chromosome Location
Chr.10: 20865333 - 20871269 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103265.1 NP_001096735.1 1 - 2 343 80
GenBank mRNA BC140558.1 - 1 - 2 343 80

View Full Product Details