Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SBSN
Entrez Gene ID
540452
Gene Name
suprabasin
Gene Aliases
-
Chromosome Location
Chr.18: 46186094 - 46190779 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104997.1 NP_001098467.1 1 - 2 1554 107
GenBank mRNA BC149806.1 - 1 - 2 1554 107

View Full Product Details