Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SH2D2A
Entrez Gene ID
530059
Gene Name
SH2 domain containing 2A
Gene Aliases
-
Chromosome Location
Chr.3: 14027673 - 14037570 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105401.2 NP_001098871.1 1 - 2 101 69
GenBank mRNA BC153239.1 - 1 - 2 102 69

View Full Product Details