Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KRT40
Entrez Gene ID
538611
Gene Name
keratin 40
Gene Aliases
-
Chromosome Location
Chr.19: 41179776 - 41186890 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105418.1 NP_001098888.1 6 - 7 1241 98
GenBank mRNA BC134653.1 - 6 - 7 1241 98

View Full Product Details