Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EWSR1
Entrez Gene ID
534073
Gene Name
EWS RNA binding protein 1
Gene Aliases
-
Chromosome Location
Chr.17: 68553309 - 68579071 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109800.2 NP_001103270.1 13 - 14 1471 153
GenBank mRNA BC153844.1 - 13 - 14 1472 153

View Full Product Details