Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAPK3
Entrez Gene ID
531391
Gene Name
mitogen-activated protein kinase 3
Gene Aliases
ERK1
Chromosome Location
Chr.25: 26179083 - 26186189 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110018.1 NP_001103488.1 8 - 9 964 69

View Full Product Details