Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AKIRIN2
Entrez Gene ID
614292
Gene Name
akirin 2
Gene Aliases
Akirin-2, C9H6ORF166
Chromosome Location
Chr.9: 62239164 - 62262032 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110087.1 NP_001103557.1 2 - 3 912 101
GenBank mRNA BC104619.1 - 2 - 3 912 101

View Full Product Details