Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SF3A2
Entrez Gene ID
789082
Gene Name
splicing factor 3a subunit 2
Gene Aliases
-
Chromosome Location
Chr.7: 21392126 - 21400800 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099211.1 NP_001092681.1 7 - 8 626 67
GenBank mRNA BC142185.1 - 7 - 8 626 67

View Full Product Details