Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
NELFA
Entrez Gene ID
789389
Gene Name
negative elongation factor complex member A
Gene Aliases
WHSC2
Chromosome Location
Chr.6: 116624401 - 116640130 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102353.1 NP_001095823.1 7 - 8 1006 115
GenBank mRNA BC112828.1 - 7 - 8 941 115
BC149974.1 - 7 - 8 1006 115

View Full Product Details