Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KHNYN
Entrez Gene ID
100124517
Gene Name
KH and NYN domain containing
Gene Aliases
-
Chromosome Location
Chr.10: 20734751 - 20744735 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102551.1 NP_001096021.1 4 - 5 1619 71
GenBank mRNA BC151400.1 - 4 - 5 1619 71

View Full Product Details