Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATAD3A
Entrez Gene ID
784353
Gene Name
ATPase family, AAA domain containing 3A
Gene Aliases
ATAD3, ATAD3B
Chromosome Location
Chr.16: 51085055 - 51101450 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105462.1 NP_001098932.1 2 - 3 309 82
GenBank mRNA BC134487.1 - 2 - 3 309 82

View Full Product Details