Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNTB2
Entrez Gene ID
786534
Gene Name
syntrophin beta 2
Gene Aliases
-
Chromosome Location
Chr.18: 36384453 - 36448425 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105466.1 NP_001098936.1 6 - 7 1557 132
GenBank mRNA BC134712.1 - 6 - 7 1557 132

View Full Product Details