Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RRP36
Entrez Gene ID
100125879
Gene Name
ribosomal RNA processing 36
Gene Aliases
C23H6orf153
Chromosome Location
Chr.23: 16661624 - 16665997 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105479.1 NP_001098949.1 6 - 7 621 64
GenBank mRNA BC148959.1 - 6 - 7 621 64

View Full Product Details