Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CGN1
Entrez Gene ID
281068
Gene Name
conglutinin
Gene Aliases
BC
Chromosome Location
Chr.28: 35391134 - 35402084 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175699.2 NP_783630.2 2 - 3 231 149
GenBank mRNA D14085.1 - 2 - 3 100 149
L18871.1 - 2 - 3 231 149
X71774.1 - - 148 149

View Full Product Details