Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PLA2G2D3
- Entrez Gene ID
-
503624
- Gene Name
- calcium-dependent phospholipase A2 PLA2G2D3
- Gene Aliases
-
-
- Chromosome Location
-
-
- Gene Symbol
- PLA2G2D1
- Entrez Gene ID
-
494318
- Gene Name
- calcium-dependent phospholipase A2 PLA2G2D1
- Gene Aliases
-
PLA2G2D2, PLA2G2D3
- Chromosome Location
-
Chr.2: 132596456 - 132609998 on Build ARS-UCD1.2
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AJ850106.1 |
- |
1 - 1 |
49 |
121 |
View Full Product Details