Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC785287
Entrez Gene ID
785287
Gene Name
ring finger protein, LIM domain interacting-like
Gene Aliases
-
Chromosome Location
Chr.X: 108854393 - 108858371 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099728.2 NP_001093198.1 2 - 2 1650 153
GenBank mRNA BC142439.1 - 2 - 2 1651 153

View Full Product Details