Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
CCDC96
Entrez Gene ID
100125235
Gene Name
coiled-coil domain containing 96
Gene Aliases
-
Chromosome Location
Chr.6: 113508435 - 113511325 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103314.1 NP_001096784.1 1 - 1 2751 146
GenBank mRNA BC149253.1 - 1 - 1 2751 146

View Full Product Details