Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
ZNF75D
Entrez Gene ID
100125581
Gene Name
zinc finger protein 75D
Gene Aliases
-
Chromosome Location
Chr.X: 18889964 - 18948896 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105039.1 NP_001098509.1 6 - 6 1751 124
GenBank mRNA BC149583.1 - 6 - 6 1751 124

View Full Product Details