Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC25A5
Entrez Gene ID
282479
Gene Name
solute carrier family 25 member 5
Gene Aliases
-
Chromosome Location
Chr.X: 3852739 - 3855413 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174659.1 NP_777084.1 4 - 4 1053 165
GenBank mRNA AB065433.1 - 4 - 4 1053 165
BC102950.1 - 4 - 4 1118 165
DQ347586.1 - 1 - 1 208 165
DQ347588.1 - 1 - 1 208 165

View Full Product Details