Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PAG2
Entrez Gene ID
337897
Gene Name
pregnancy-associated glycoprotein 2
Gene Aliases
-
Chromosome Location
Chr.29: 37572051 - 37581465 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_176614.1 NP_788787.1 9 - 9 1043 127
GenBank mRNA AB098772.1 - 1 - 1 241 127
AB098799.1 - 1 - 1 254 127
AB098821.1 - 1 - 1 253 127
AB098830.1 - 1 - 1 256 127
AB098856.1 - 1 - 1 254 127
AB098905.1 - - 231 127
AY911498.1 - 9 - 9 1043 127
BC149444.1 - 9 - 9 1084 127
L06151.1 - 9 - 9 1043 127

View Full Product Details