Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TCRA
Entrez Gene ID
785621
Gene Name
T cell receptor, alpha
Gene Aliases
TRA@
Chromosome Location
Chr.10: 22731266 - 22732204 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC134751.1 - - 250 136

View Full Product Details