Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHTF8
Entrez Gene ID
618186
Gene Name
chromosome transmission fidelity factor 8
Gene Aliases
CTF8, DERPC
Chromosome Location
Chr.18: 36332461 - 36343429 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114969.2 NP_001108441.1 3 - 4 255 91
GenBank mRNA EV621490.1 - 3 - 4 263 91

View Full Product Details