Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HBEGF
Entrez Gene ID
522921
Gene Name
heparin binding EGF like growth factor
Gene Aliases
DTR, HB-EGF
Chromosome Location
Chr.7: 51533600 - 51544783 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144090.1 NP_001137562.1 1 - 2 329 58

View Full Product Details