Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KEAP1
Entrez Gene ID
532791
Gene Name
kelch like ECH associated protein 1
Gene Aliases
-
Chromosome Location
Chr.7: 15009368 - 15018662 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101142.1 NP_001094612.1 4 - 5 1623 72
GenBank mRNA BC151545.1 - 4 - 5 1623 72

View Full Product Details