The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001123365.1 | NP_001116837.1 | 5 - 6 | 1012 | 66 |
GenBank mRNA | EU527855.1 | - | 5 - 6 | 1012 | 66 |