Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPACA4
Entrez Gene ID
767953
Gene Name
sperm acrosome associated 4
Gene Aliases
-
Chromosome Location
Chr.18: 55252155 - 55253025 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077023.2 NP_001070491.1 1 - 1 149 73
GenBank mRNA BC108186.1 - 1 - 1 323 73

View Full Product Details