Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CLDN8
Entrez Gene ID
538761
Gene Name
claudin 8
Gene Aliases
-
Chromosome Location
Chr.1: 5770225 - 5772220 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098096.1 NP_001091565.1 1 - 1 824 77
GenBank mRNA BC134660.1 - 1 - 1 824 77

View Full Product Details