Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZNF35
Entrez Gene ID
782819
Gene Name
zinc finger protein 35
Gene Aliases
-
Chromosome Location
Chr.22: 16448663 - 16461755 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205425.3 NP_001192354.3 2 - 3 351 65
GenBank mRNA BC149195.1 - 2 - 3 368 65

View Full Product Details