Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FBXO48
Entrez Gene ID
786890
Gene Name
F-box protein 48
Gene Aliases
-
Chromosome Location
Chr.11: 66859773 - 66865138 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205456.1 NP_001192385.1 1 - 2 305 90

View Full Product Details